Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130912.2 | NP_001124384.1 | 7 | 696 | ||
NM_001320831.1 | NP_001307760.1 | 7 | 728 | |||
NM_006704.4 | NP_006695.1 | 6 | 600 | |||
XM_011534891.2 | XP_011533193.1 | 7 | 791 | |||
XM_011534893.2 | XP_011533195.1 | 6 | 509 | |||
XM_017020365.1 | XP_016875854.1 | 7 | 681 | |||
XM_017020366.1 | XP_016875855.1 | 7 | 556 | |||
XM_017020367.1 | XP_016875856.1 | 7 | 590 | |||
XM_017020368.1 | XP_016875857.1 | 7 | 602 | |||
GenBank mRNA | AF132856.1 | - | 6 | 370 | ||
AJ344097.1 | - | 6 | 463 | |||
AK291402.1 | - | 7 | 513 | |||
AK292128.1 | - | 6 | 460 | |||
AK302363.1 | - | 7 | 575 | |||
AY271314.1 | - | 7 | 533 | |||
AY321358.1 | - | 7 | 548 | |||
BC000911.1 | - | 6 | 425 | 2985858 | ||
BT009798.1 | - | 6 | 370 |
Gene Symbol: SUGT1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003667.3 | 5 | 756 | |||
GenBank mRNA | DA804368.1 | - | 5 | 441 |