Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025907.3 | NP_080183.1 | 6 | 1797 | ||
GenBank mRNA | AK082949.1 | - | 6 | 1854 | ||
BC030449.1 | - | 6 | 1798 | 5400506 |