Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025910.3 | NP_080186.3 | 5 | 966 | ||
GenBank mRNA | AB177385.1 | - | 4 | 767 | ||
AK007810.1 | - | 5 | 936 | |||
AK013451.1 | - | 5 | 926 | |||
AK014426.1 | - | 5 | 913 | |||
AK031671.1 | - | 5 | 900 | |||
AK219613.1 | - | 2 | 122 | |||
BC023462.1 | - | 5 | 1221 | 5041558 | ||
BC025109.1 | - | 5 | 888 | 4236994 | ||
BC058242.1 | - | 5 | 875 | 6308890 |