Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025910.3 | NP_080186.3 | 9, 10 | 1437 | ||
GenBank mRNA | AB177385.1 | - | 8, 9 | 1238 | ||
AK007810.1 | - | 9, 10 | 1407 | |||
AK013451.1 | - | 9, 10 | 1397 | |||
AK031671.1 | - | 9, 10 | 1371 | |||
BC023462.1 | - | 9, 10 | 1692 | 5041558 | ||
BC025109.1 | - | 9, 10 | 1359 | 4236994 | ||
BC058242.1 | - | 9, 10 | 1346 | 6308890 |