Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC023462.1 | - | 10 | 1765 | 5041558 | |
BC025109.1 | - | 10 | 1432 | 4236994 |