Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277273.1 | NP_001264202.1 | 13 | 1167 | ||
NM_025923.3 | NP_080199.1 | 13 | 1182 | |||
GenBank mRNA | AF513619.1 | - | 13 | 1182 | ||
AK008605.1 | - | 13 | 1068 | |||
AK011520.1 | - | 13 | 1087 | |||
AK011752.1 | - | 13 | 1059 | |||
AK011903.1 | - | 13 | 1050 | |||
AK017431.1 | - | 13 | 1062 | |||
AK018018.1 | - | 13 | 1064 | |||
AK076017.1 | - | 13 | 1065 | |||
AK168538.1 | - | 13 | 1066 | |||
BC032876.1 | - | 13 | 1056 | 1282316 | ||
BC066181.1 | - | 13 | 1111 | 6435171 | ||
CN455644.1 | - | 10 | 735 |