Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025927.4 | NP_080203.1 | 2, 3 | 410 | ||
GenBank mRNA | AK011163.1 | - | 2, 3 | 279 | ||
AK011605.1 | - | 2, 3 | 269 | |||
AK081922.1 | - | 2, 3 | 271 | |||
AK157771.1 | - | 2, 3 | 272 | |||
BC020130.1 | - | 2, 3 | 255 | 3990098 |