Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025937.4 | NP_080213.3 | 6 | 1035 | ||
GenBank mRNA | AK011492.1 | - | 6 | 986 | ||
AK038344.1 | - | 6 | 983 | |||
AK089943.1 | - | 6 | 963 | |||
AY388959.1 | - | 6 | 986 | |||
BC026774.1 | - | 6 | 1003 | 4019392 |