Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025948.2 | NP_080224.1 | 4 | 518 | ||
GenBank mRNA | AK003611.1 | - | 2 | 78 | ||
AK012276.1 | - | 3 | 281 | |||
AK182421.1 | - | 2 | 185 | |||
AK186175.1 | - | 1 | 38 | |||
AK189673.1 | - | 3 | 253 | |||
AK205955.1 | - | 2 | 186 | |||
BC031521.1 | - | 4 | 518 | 2655161 |