Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006571.3 | NP_006562.1 | 7 | 801 | ||
GenBank mRNA | BC013175.1 | - | 7 | 745 | 3925231 | |
D84145.1 | - | 7 | 801 |