Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025975.5 | NP_080251.3 | 5 | 1463 | ||
GenBank mRNA | AK010186.1 | - | 5 | 1397 | ||
AK088277.1 | - | 5 | 1464 | |||
AK134558.1 | - | 826 | ||||
BC030032.1 | - | 5 | 1390 | 1332100 |