Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025980.2 | NP_080256.2 | 1 | 1150 | ||
GenBank mRNA | AK012426.1 | - | 1 | 886 | ||
AY046077.1 | - | 1 | 1150 | |||
BC020293.1 | - | 1 | 1147 | |||
BC048088.1 | - | 2 | 619 | |||
BC069891.1 | - | 1 | 903 | 5391567 |