Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025980.2 | NP_080256.2 | 1 | 2432 | ||
GenBank mRNA | AK004508.1 | - | 1 | 637 | ||
AK012426.1 | - | 1 | 2168 | |||
AY046077.1 | - | 1 | 2432 | |||
BC048088.1 | - | 2 | 1901 | |||
BC069891.1 | - | 1 | 2185 | 5391567 |