Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025990.4 | NP_080266.2 | 4 | 490 | ||
GenBank mRNA | AK009329.1 | - | 4 | 486 | ||
AK009441.1 | - | 4 | 492 | |||
AK009531.1 | - | 4 | 486 | |||
AK009562.1 | - | 4 | 486 | |||
AK009580.1 | - | 4 | 490 | |||
AK009629.1 | - | 4 | 489 | |||
AK009695.1 | - | 4 | 487 | |||
AK009803.1 | - | 4 | 488 | |||
AK009835.1 | - | 4 | 490 | |||
AK010012.1 | - | 4 | 490 | |||
AK010051.1 | - | 4 | 487 | |||
AK010115.1 | - | 4 | 486 | |||
AK010143.1 | - | 3 | 432 | |||
AK010145.1 | - | 4 | 490 | |||
AK010158.1 | - | 4 | 486 | |||
AK010159.1 | - | 4 | 489 | |||
BC115526.1 | - | 4 | 469 | 40101263 | ||
BC115527.1 | - | 4 | 466 | 40101264 |