Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006711244.3 | XP_006711307.1 | 7 | 2982 |
Gene Symbol: CCT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282907.1 | NP_001269836.1 | 16 | 2199 | ||
NM_001282908.1 | NP_001269837.1 | 15 | 1916 | |||
NM_001282909.1 | NP_001269838.1 | 14 | 1811 | |||
NM_006585.3 | NP_006576.2 | 15 | 1891 | |||
GenBank mRNA | AK293705.1 | - | 16 | 1753 | ||
AK298801.1 | - | 14 | 1665 | |||
AK308063.1 | - | 15 | 1743 | |||
BC012584.1 | - | 15 | 1758 | 4133182 | ||
BC035472.1 | - | 15 | 1707 | |||
BC072001.1 | - | 15 | 1749 | 6298609 | ||
BC095470.1 | - | 15 | 1731 | 30523886 | ||
D13627.1 | - | 15 | 1712 |