Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026014.3 | NP_080290.3 | 2 | 379 | ||
GenBank mRNA | AB086655.1 | - | 2 | 358 | ||
AF477481.1 | - | 2 | 379 | |||
AK012041.1 | - | 2 | 356 | |||
AK017609.1 | - | 2 | 353 | |||
AK028287.1 | - | 2 | 351 | |||
AK049163.1 | - | 2 | 351 | |||
AK076527.1 | - | 2 | 368 | |||
AK083015.1 | - | 2 | 354 | |||
AK185323.1 | - | 1 | 146 | |||
AK208949.1 | - | 1 | 88 | |||
BC013842.1 | - | 2 | 334 | |||
BC048076.1 | - | 2 | 379 | 6307476 |