Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026014.3 | NP_080290.3 | 10 | 1936 | ||
GenBank mRNA | AB086655.1 | - | 10 | 1915 | ||
AF477481.1 | - | 10 | 1936 | |||
AK012041.1 | - | 10 | 1913 | |||
AK017609.1 | - | 10 | 1910 | |||
AK021216.1 | - | 1 | 361 | |||
AK028287.1 | - | 10 | 1908 | |||
AK049163.1 | - | 10 | 1908 | |||
AK076527.1 | - | 9 | 2018 | |||
AK083015.1 | - | 10 | 1911 | |||
AK184088.1 | - | 1 | 155 | |||
AK192601.1 | - | 1 | 243 | |||
AK192707.1 | - | 1 | 243 | |||
AK206789.1 | - | 1 | 127 | |||
AK213623.1 | - | 1 | 243 | |||
AK215658.1 | - | 1 | 115 | |||
AK218896.1 | - | 1 | 151 | |||
BC013842.1 | - | 10 | 1891 | |||
BC024485.1 | - | 7 | 1187 | |||
BC048076.1 | - | 10 | 1936 | 6307476 |