Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026545.3 | NP_080821.3 | 7 | 1080 | ||
GenBank mRNA | AK002921.1 | - | 7 | 919 | ||
AK003436.1 | - | 7 | 921 | |||
AK020132.1 | - | 7 | 939 | |||
AK027973.1 | - | 7 | 884 | |||
AK051257.1 | - | 7 | 932 | |||
AK150493.1 | - | 7 | 923 | |||
AK150982.1 | - | 7 | 887 | |||
AK153329.1 | - | 7 | 921 | |||
AK159216.1 | - | 7 | 889 | |||
AK159795.1 | - | 7 | 905 | |||
AK159926.1 | - | 7 | 915 | |||
AK166718.1 | - | 7 | 914 | |||
AK167948.1 | - | 7 | 889 | |||
AK168886.1 | - | 6 | 1551 | |||
BC004075.1 | - | 7 | 898 | 3591796 | ||
BC005717.1 | - | 7 | 808 | 2609781 |