Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026545.3 | NP_080821.3 | 1 | 363 | ||
GenBank mRNA | AK002921.1 | - | 1 | 202 | ||
AK003436.1 | - | 1 | 204 | |||
AK020132.1 | - | 1 | 222 | |||
AK027973.1 | - | 1 | 167 | |||
AK051257.1 | - | 1 | 215 | |||
AK150493.1 | - | 1 | 206 | |||
AK150982.1 | - | 1 | 170 | |||
AK153329.1 | - | 1 | 204 | |||
AK159216.1 | - | 1 | 172 | |||
AK166718.1 | - | 1 | 197 | |||
AK168886.1 | - | 1 | 171 | |||
CF732286.1 | - | 1 | 363 |