Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278585.1 | NP_001265514.1 | 10 | 1539 | ||
NM_001278586.1 | NP_001265515.1 | 11 | 1740 | |||
NM_006587.3 | NP_006578.2 | 12 | 1851 | |||
GenBank mRNA | AF133845.1 | - | 12 | 1787 | ||
AK297675.1 | - | 11 | 1701 | |||
AK302816.1 | - | 11 | 1526 | |||
AK304038.1 | - | 11 | 1616 | |||
AK304466.1 | - | 11 | 1740 | |||
BC110451.2 | - | 12 | 1787 | 40027293 |