Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028260.2 | NP_082536.1 | 6 | 627 | ||
GenBank mRNA | AK005356.1 | - | 6 | 628 | ||
AK009570.1 | - | 6 | 585 | |||
AK010075.1 | - | 6 | 585 | |||
AK011278.1 | - | 6 | 628 | |||
AK011382.1 | - | 6 | 628 | |||
AK011420.1 | - | 6 | 589 | |||
AK011466.1 | - | 6 | 600 | |||
AK011618.1 | - | 6 | 589 | |||
AK011872.1 | - | 6 | 591 | |||
AK012181.1 | - | 580 | ||||
AK015978.1 | - | 6 | 598 | |||
AK078300.1 | - | 6 | 614 | |||
AK135670.1 | - | 6 | 587 | |||
AK168202.1 | - | 6 | 576 | |||
BC008259.1 | - | 6 | 586 | 2649406 | ||
BC081433.1 | - | 5 | 470 | 6814816 |