Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029979.3 | NP_084255.2 | 6 | 1436 | ||
GenBank mRNA | AB060155.1 | - | 6 | 1380 | ||
AF145374.1 | - | 6 | 1420 | |||
AK003000.1 | - | 6 | 1395 | |||
AK032635.1 | - | 6 | 1409 | |||
AK075877.1 | - | 6 | 1407 | |||
AK146541.1 | - | 5 | 887 | |||
AK188346.1 | - | 1 | 122 | |||
AK210014.1 | - | 1 | 189 | |||
BC049105.1 | - | 6 | 1386 | 6418046 | ||
BC053494.1 | - | 6 | 1301 | |||
BC145783.1 | - | 6 | 1430 | 40131086 | ||
BC145809.1 | - | 6 | 1430 | 40131112 |