Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256725.1 | NP_001243654.1 | 8 | 1173 | ||
NM_001256726.1 | NP_001243655.1 | 8 | 1173 | |||
NM_001256727.1 | NP_001243656.1 | 8 | 1058 | |||
NM_001256728.1 | NP_001243657.1 | 7 | 983 | |||
NM_006590.3 | NP_006581.2 | 8 | 1173 | |||
NR_046347.1 | 8 | 1061 | ||||
XM_006711922.1 | XP_006711985.1 | 8 | 1041 | |||
XM_006711923.2 | XP_006711986.1 | 8 | 1120 | |||
XM_011532487.1 | XP_011530789.1 | 8 | 1126 | |||
XM_011532488.1 | XP_011530790.1 | 8 | 1059 | |||
XM_017003182.1 | XP_016858671.1 | 8 | 1130 | |||
GenBank mRNA | AF130096.1 | - | 8 | 762 | ||
AF132955.1 | - | 8 | 1078 | |||
AF161450.1 | - | 3 | 159 | |||
AF353989.1 | - | 8 | 1080 | |||
AK001047.1 | - | 8 | 1071 | |||
AK001772.1 | - | 4 | 466 | |||
AK056334.1 | - | 8 | 1042 | |||
AK129872.1 | - | 9 | 1054 | |||
AK222852.1 | - | 8 | 1076 | |||
AK289451.1 | - | 8 | 1127 | |||
AK295257.1 | - | 7 | 981 | |||
AK302236.1 | - | 8 | 1035 | |||
AK309485.1 | - | 10 | 976 | |||
AL117416.1 | - | 6 | 740 | |||
BC001384.2 | - | 8 | 1062 | |||
BC067273.1 | - | 8 | 1101 | 6503509 |