Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290437.1 | NP_001277366.1 | 2 | 954 | ||
NM_033134.3 | NP_149125.1 | 1 | 1238 | |||
GenBank mRNA | AF226683.1 | - | 1 | 1200 | ||
AK047245.1 | - | 1 | 1223 | |||
AK080075.1 | - | 1 | 1220 | |||
AK154097.1 | - | 1 | 1239 | |||
AK158135.1 | - | 1 | 1223 | |||
AK170174.1 | - | 1 | 1171 | |||
AK170786.1 | - | 1 | 1195 | |||
AK170812.1 | - | 1 | 1209 | |||
BC052717.1 | - | 1 | 1207 | 6837339 | ||
BC057566.1 | - | 1 | 1203 | |||
BC080295.1 | - | 1 | 1203 | 30354397 |