Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133676.2 | NP_598437.2 | 11 | 1224 | ||
GenBank mRNA | AK005291.1 | - | 11 | 1173 | ||
AK049480.1 | - | 9 | 1712 | |||
AK076246.1 | - | 11 | 1184 | |||
AK142730.1 | - | 11 | 1224 | |||
AK145181.1 | - | 11 | 1152 | |||
AK157554.1 | - | 1175 | ||||
AK166415.1 | - | 11 | 1184 | |||
AK167893.1 | - | 11 | 1165 | |||
AK207563.1 | - | 3 | 290 | |||
BC002296.1 | - | 11 | 1159 | 3590664 | ||
BC091757.1 | - | 11 | 1146 | 4925017 |