Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026994.1 | NP_081270.1 | 6 | 562 | ||
NM_133679.2 | NP_598440.1 | 7 | 607 | |||
GenBank mRNA | AK008846.1 | - | 7 | 383 | ||
AK010433.1 | - | 7 | 411 | |||
AK017995.1 | - | 6 | 563 | |||
AK145827.1 | - | 7 | 431 | |||
BC010479.1 | - | 7 | 404 | 3968727 | ||
BC019387.1 | - | 6 | 377 | 4164847 | ||
BY717916.1 | - | 6 | 563 |