Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083901.1 | NP_001077370.1 | 12 | 1465 | ||
NM_133680.3 | NP_598441.3 | 11 | 1562 | |||
GenBank mRNA | AK146596.1 | - | 12 | 1458 | ||
AK190739.1 | - | 1 | 36 | |||
BC018395.1 | - | 4 | 350 | |||
BC033469.1 | - | 11 | 1524 | 4951631 |