Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133795.1 | NP_598556.1 | 6, 7 | 775 | ||
GenBank mRNA | AK077715.1 | - | 6, 7 | 788 | ||
AK166431.1 | - | 6, 7 | 782 | |||
AK170032.1 | - | 6, 7 | 786 | |||
AK170230.1 | - | 6, 7 | 785 | |||
BC010236.1 | - | 6, 7 | 775 | 3581904 |