Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133934.4 | NP_598695.1 | 7 | 467 | ||
GenBank mRNA | AK017996.1 | - | 7 | 384 | ||
AK173116.1 | - | 435 | ||||
BC020103.1 | - | 7 | 431 | 3673446 | ||
BC037695.1 | - | 7 | 472 | 4009615 |