Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252444.1 | NP_001239373.1 | 21 | 1855 | ||
NM_144954.3 | NP_659203.1 | 21 | 1757 | |||
GenBank mRNA | AK012472.1 | - | 21 | 1836 | ||
BC028899.1 | - | 21 | 1718 | 4021590 |