Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcamr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170632.1 | NP_001164103.1 | 5 | 1147 | ||
NM_144960.2 | NP_659209.2 | 5 | 979 | |||
GenBank mRNA | AB048834.1 | - | 5 | 991 | ||
BC110549.1 | - | 5 | 1150 | 40047147 | ||
BC110550.1 | - | 5 | 982 | 40047153 | ||
FJ011703.1 | - | 4 | 1060 |