Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145066.4 | NP_659503.1 | 4 | 1777 | ||
GenBank mRNA | AF254416.2 | - | 1 | 813 | ||
AK043090.1 | - | 2 | 1002 | |||
AK047740.1 | - | 3 | 980 | |||
BC026975.1 | - | 1 | 994 | 5363257 | ||
BC036271.1 | - | 1 | 994 | |||
BC065154.1 | - | 4 | 1777 | 6418997 |
Set Membership: |
G-protein coupled receptor |