Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145568.3 | NP_663543.1 | 4 | 1079 | ||
GenBank mRNA | AK146818.1 | - | 3 | 1018 | ||
AK162678.1 | - | 3 | 1007 | |||
BC005704.1 | - | 2 | 724 | 3598565 | ||
BC046798.1 | - | 3 | 975 | 6403577 | ||
BC069863.1 | - | 4 | 1079 | 6306896 |