Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145944.4 | NP_666056.1 | 5, 6 | 392 | ||
GenBank mRNA | AK012172.1 | - | 5, 6 | 340 | ||
AK016286.1 | - | 5, 6 | 304 | |||
AK078345.1 | - | 5, 6 | 393 | |||
AK089982.1 | - | 5, 6 | 317 | |||
AK139180.1 | - | 4, 5 | 2768 | |||
BC025545.1 | - | 340 | ||||
BC128015.1 | - | 5, 6 | 284 | 40136066 | ||
BC148235.1 | - | 5, 6 | 291 | 40136084 |