Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145944.4 | NP_666056.1 | 7 | 557 | ||
GenBank mRNA | AK012172.1 | - | 7 | 505 | ||
AK016286.1 | - | 7 | 469 | |||
AK078345.1 | - | 7 | 558 | |||
AK089982.1 | - | 7 | 482 | |||
AK139180.1 | - | 6 | 2933 | |||
BC025545.1 | - | 505 | ||||
BC128015.1 | - | 7 | 449 | 40136066 | ||
BC148235.1 | - | 7 | 456 | 40136084 |