Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sv2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109753.1 | NP_001103223.1 | 13 | 3182 | ||
NM_153579.4 | NP_705807.2 | 13 | 3389 | |||
GenBank mRNA | AK029736.1 | - | 14 | 3214 | ||
AK043830.1 | - | 13 | 3109 | |||
AK122359.1 | - | 12 | 2334 | |||
AK139478.1 | - | 13 | 3084 | |||
AK140104.1 | - | 13 | 3103 | |||
AK147261.1 | - | 13 | 3176 | |||
AK159028.1 | - | 13 | 3088 | |||
BC034651.1 | - | 3 | 1118 | |||
BC060224.1 | - | 12 | 2770 | 30356990 | ||
BC079568.1 | - | 13 | 3388 |