Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310022A10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175107.5 | NP_780316.3 | 10 | 1261 | ||
GenBank mRNA | BC023369.1 | - | 10 | 1240 | 5038597 | |
BE854222.1 | - | 1 | 71 |