Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175113.3 | NP_780322.2 | 9 | 1312 | ||
GenBank mRNA | AK014359.1 | - | 9 | 1428 | ||
AK028755.1 | - | 9 | 1312 | |||
AK129300.1 | - | 9 | 1279 | |||
AK164396.1 | - | 9 | 1245 | |||
AK165976.1 | - | 9 | 1264 | |||
AK167222.1 | - | 9 | 1278 | |||
BC049083.1 | - | 5 | 1440 | |||
BC052648.1 | - | 9 | 1230 | 30030801 |