Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145158.1 | NP_001138630.1 | 17 | 7010 | ||
NM_006620.3 | NP_006611.1 | 18 | 7136 | |||
GenBank mRNA | AJ459826.1 | - | 18 | 7125 |