Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113532.1 | NP_001107004.1 | 5 | 349 | ||
NM_001113533.1 | NP_001107005.1 | 5 | 349 | |||
NM_175394.2 | NP_780603.1 | 5 | 397 | |||
GenBank mRNA | AJ276707.1 | - | 3 | 107 | ||
AK031965.1 | - | 5 | 405 | |||
AK076111.1 | - | 5 | 391 | |||
AK156334.1 | - | 5 | 245 | |||
BC046416.1 | - | 5 | 278 | |||
BC047338.1 | - | 5 | 278 | |||
BC093504.1 | - | 5 | 282 | |||
BU516500.1 | - | 484 |