Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113532.1 | NP_001107004.1 | 6 | 791 | ||
NM_175394.2 | NP_780603.1 | 6 | 839 | |||
GenBank mRNA | AK031965.1 | - | 6 | 848 | ||
AK076111.1 | - | 6 | 833 | |||
AK220249.1 | - | 2 | 446 | |||
BC046416.1 | - | 6 | 720 | |||
BC047338.1 | - | 6 | 720 | |||
BC093504.1 | - | 6 | 724 |