Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp280b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177475.3 | NP_803426.2 | 2 | 2819 | ||
GenBank mRNA | AK031331.1 | - | 2 | 2717 | ||
AK049339.1 | - | 1 | 1660 | |||
AK079824.1 | - | 1 | 484 | |||
AK144644.1 | - | 2 | 2777 | |||
AK144736.1 | - | 2 | 2777 | |||
AK157463.1 | - | 1 | 2713 | |||
BC094574.1 | - | 2 | 2748 | 6856157 | ||
BC117873.1 | - | 3 | 2381 | |||
BC117874.2 | - | 2 | 2705 | 40095593 |