Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gskip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178613.3 | NP_848728.2 | 2 | 334 | ||
GenBank mRNA | AK017049.1 | - | 858 | |||
AK020785.1 | - | 2 | 272 | |||
AK029484.1 | - | 2 | 246 | |||
AK042664.1 | - | 2 | 295 | |||
AK079775.1 | - | 2 | 280 | |||
AK168962.1 | - | 2 | 298 | |||
AK171097.1 | - | 2 | 311 | |||
BC031494.1 | - | 2 | 302 | 3156862 |