Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190322.2 | NP_001177251.1 | 5 | 602 | ||
NM_001190323.2 | NP_001177252.1 | 5 | 478 | |||
NM_001190324.2 | NP_001177253.1 | 5 | 433 | |||
NM_001285804.1 | NP_001272733.1 | 5 | 633 | |||
NM_181391.4 | NP_852056.1 | 5 | 647 | |||
GenBank mRNA | AK003308.1 | - | 5 | 479 | ||
AK007844.1 | - | 5 | 622 | |||
AK145105.1 | - | 5 | 622 | |||
AK168647.1 | - | 5 | 577 | |||
AK169018.1 | - | 6 | 661 | |||
AV042014.2 | - | 3 | 242 | |||
BC030366.1 | - | 5 | 605 | 5370748 | ||
BX527959.1 | - | 5 | 394 | |||
BY578448.1 | - | 4 | 365 | |||
CB057699.1 | - | 5 | 559 |