Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173547.2 | NP_001167018.1 | 10 | 1655 | ||
NM_001242615.2 | NP_001229544.1 | 9 | 1602 | |||
NM_001242616.2 | NP_001229545.1 | 9 | 1599 | |||
NM_001290738.1 | NP_001277667.1 | 8 | 1521 | |||
NM_181516.6 | NP_852657.1 | 10 | 1652 | |||
GenBank mRNA | AK004035.1 | - | 10 | 1607 | ||
AK075615.1 | - | 10 | 1609 | |||
AK076481.1 | - | 10 | 1618 | |||
AK160327.1 | - | 9 | 1568 | |||
AK170662.1 | - | 9 | 2194 | |||
AK211183.1 | - | 1 | 210 | |||
BC015305.1 | - | 10 | 1617 | 4221041 | ||
BC064023.1 | - | 1 | 1063 |