Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146211.1 | NP_001139683.1 | 4 | 216 | ||
NM_001146212.1 | NP_001139684.1 | 4 | 219 | |||
NM_183086.2 | NP_898909.2 | 4 | 309 | |||
GenBank mRNA | AK004151.1 | - | 4 | 213 | ||
AK142675.1 | - | 3 | 3956 | |||
BC048912.1 | - | 4 | 312 | 4238014 | ||
BC096043.1 | - | 3 | 174 | |||
BC096657.1 | - | 3 | 124 | 5144028 | ||
DV046794.1 | - | 4 | 230 |