Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC048912.1 | - | 7 | 712 | 4238014 | |
BC096657.1 | - | 6 | 524 | 5144028 |