Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164274.1 | NP_001157746.1 | 2 | 690 | ||
NM_013720.2 | NP_038748.2 | 2 | 690 | |||
GenBank mRNA | AF205935.1 | - | 1 | 504 | ||
AK032570.1 | - | 2 | 671 | |||
AK046252.1 | - | 2 | 691 | |||
AK133775.1 | - | 2 | 907 | |||
AK139919.1 | - | 2 | 642 | |||
AK145491.1 | - | 2 | 639 | |||
AK162302.1 | - | 2 | 687 | |||
AK172962.1 | - | 2 | 613 |