Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naglu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013792.2 | NP_038820.2 | 6 | 1819 | ||
GenBank mRNA | AK009286.1 | - | 1 | 389 | ||
AK143546.1 | - | 6 | 1821 | |||
AK158774.1 | - | 4 | 2443 | |||
AK164125.1 | - | 6 | 1798 | |||
AK170758.1 | - | 6 | 1802 | |||
AK178593.1 | - | 1 | 52 | |||
AK208064.1 | - | 1 | 91 | |||
BC055733.1 | - | 6 | 1793 | 6411530 | ||
U85247.1 | - | 6 | 1788 |