Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nagpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013796.3 | NP_038824.2 | 2 | 561 | ||
GenBank mRNA | AK032158.1 | - | 2 | 562 | ||
AK087468.1 | - | 2 | 561 | |||
AK138035.1 | - | 2 | 554 | |||
AK155450.1 | - | 2 | 540 | |||
AK159629.1 | - | 2 | 540 | |||
AK162633.1 | - | 2 | 543 | |||
AK168830.1 | - | 1 | 648 | |||
AK169539.1 | - | 1 | 637 | |||
AK170219.1 | - | 2 | 543 | |||
AK211874.1 | - | 1 | 76 | |||
BC019640.1 | - | 1 | 623 | |||
BC039790.1 | - | 2 | 553 | 4951805 |